Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297746.1 | NP_001284675.1 | 4 | 2307 | |
NM_016076.4 | NP_057160.2 | 5 | 2406 | ||
NR_123735.1 | 5 | 2401 | |||
NR_123736.1 | 4 | 2312 | |||
XM_011544203.2 | XP_011542505.1 | 6 | 2491 | ||
XM_011544205.2 | XP_011542507.1 | 5 | 3057 | ||
XM_011544206.2 | XP_011542508.1 | 5 | 2130 | ||
XM_011544207.2 | XP_011542509.1 | 4 | 3497 | ||
GenBank mRNA | AL049397.1 | - | 1 | 33 | |
BG290656.1 | - | 286 |