Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320238.1 | NP_001307167.1 | 13 | 2048 | |
NM_001320239.1 | NP_001307168.1 | 13 | 1992 | ||
NM_001320240.1 | NP_001307169.1 | 17 | 2523 | ||
NM_020850.2 | NP_065901.1 | 14 | 2070 | ||
XM_005256070.2 | XP_005256127.1 | 14 | 2068 | ||
XM_005256072.2 | XP_005256129.1 | 13 | 2015 | ||
XM_006721238.3 | XP_006721301.1 | 9 | 1825 | ||
XM_011523245.2 | XP_011521547.1 | 11 | 1807 | ||
XM_011523247.2 | XP_011521549.1 | 11 | 1613 | ||
XM_017023507.1 | XP_016878996.1 | 11 | 1584 | ||
XM_017023508.1 | XP_016878997.1 | 15 | 2228 | ||
XM_017023509.1 | XP_016878998.1 | 12 | 1573 | ||
GenBank mRNA | AB040897.1 | - | 14 | 1959 | |
AK055727.1 | - | 17 | 2428 | ||
AK295530.1 | - | 13 | 1956 | ||
AK298806.1 | - | 13 | 1919 | ||
AY337313.1 | - | 14 | 2070 | ||
BC099917.1 | - | 14 | 1975 | 30528481 | |
BC121176.1 | - | 14 | 2068 | 40122561 | |
BC121177.2 | - | 14 | 2068 | 40122562 |