Gene Transcripts

Gene Symbol
CHIA
Entrez Gene ID
27159
Gene Name
chitinase, acidic
Gene Aliases
AMCASE, CHIT2, TSA1902
Chromosome Location
Chr. 1: 111290852 - 111320566 on Build GRCh38
UniGene ID
Hs.128814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040623.2 NP_001035713.1 7 576
NM_001258001.1 NP_001244930.1 8 742
NM_001258002.1 NP_001244931.1 5 453
NM_001258003.1 NP_001244932.1 7 649
NM_001258004.1 NP_001244933.1 9 881
NM_001258005.1 NP_001244934.1 8 715
NM_021797.3 NP_068569.2 6 619
NM_201653.3 NP_970615.2 9 944
XM_006710577.3 XP_006710640.1 6 611
XM_017001047.1 XP_016856536.1 4 464
XM_017001048.1 XP_016856537.1 7 652
GenBank mRNA AB025008.1 - 6 609
AB025009.1 - 5 443
AF290004.1 - 9 880
AK098814.1 - 5 3500
AK290702.1 - 8 744
AY789444.1 - 8 678
AY789445.1 - 6 478
AY825504.1 - 7 639
AY911310.1 - 8 737
AY911311.1 - 7 551
BC036339.1 - 6 557 5184463
BC047336.1 - 7 566 5185486
BC106910.1 - 5 525 40031872
BC139901.1 - 7 571
BC139920.1 - 7 571

View Full Product Details