Gene Transcripts
- Gene Symbol
- SPOCD1
- Entrez Gene ID
- 90853
- Gene Name
- SPOC domain containing 1
- Gene Aliases
- PPP1R146
- Chromosome Location
- Chr. 1: 31790422 - 31816051 on Build GRCh38
- UniGene ID
- Hs.62604
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281987.1 |
NP_001268916.1
|
7, 8 |
1992 |
|
NM_001281988.1 |
NP_001268917.1
|
6, 7 |
570 |
|
NM_144569.5 |
NP_653170.3
|
7, 8 |
1992 |
|
XM_011542413.2 |
XP_011540715.1
|
7, 8 |
561 |
|
XM_011542414.2 |
XP_011540716.1
|
6, 7 |
756 |
|
XM_017002779.1 |
XP_016858268.1
|
6, 7 |
1901 |
|
XM_017002780.1 |
XP_016858269.1
|
5, 6 |
1859 |
|
XM_017002781.1 |
XP_016858270.1
|
6, 7 |
1901 |
|
XM_017002782.1 |
XP_016858271.1
|
6, 7 |
1130 |
|
GenBank mRNA |
AK091016.1 |
- |
6, 7 |
541 |
|
AK131447.1 |
- |
7, 8 |
1921 |
|
BC114430.1 |
- |
7, 8 |
1918 |
40038407 |
View Full Product Details