Gene Transcripts

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
Chromosome Location
Chr. 1: 108815898 - 108858242 on Build GRCh38
UniGene ID
Hs.729441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152763.4 NP_689976.2 6 1628
NR_049760.1 7 1769
GenBank mRNA AK095517.1 - 7 1755
AL832216.1 - 6 1618
BC030647.2 - 6 1625
BC150642.1 - 5 1383 9057012

View Full Product Details