Gene Transcripts

Gene Symbol
LAYN
Entrez Gene ID
143903
Gene Name
layilin
Gene Aliases
Chromosome Location
Chr. 11: 111540280 - 111563464 on Build GRCh38
UniGene ID
Hs.503831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258390.1 NP_001245319.1 8 1308
NM_001258391.1 NP_001245320.1 6 1126
NM_001318799.1 NP_001305728.1 7 918
NM_178834.4 NP_849156.1 7 1284
XM_006718769.3 XP_006718832.1 7 897
XM_006718770.3 XP_006718833.1 6 954
GenBank mRNA AK055539.1 - 7 1285
AK055654.1 - 7 1072
AK293317.1 - 7 1115
AK296231.1 - 6 914
AK298165.1 - 8 943
AY358554.1 - 8 1133
BC025407.1 - 7 1077 4820414

View Full Product Details