Gene Transcripts

Gene Symbol
PDDC1
Entrez Gene ID
347862
Gene Name
Parkinson disease 7 domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 767095 - 777502 on Build GRCh38
UniGene ID
Hs.218362
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318818.1 NP_001305747.1 7 1249
NM_001318820.1 NP_001305749.1 6 1307
NM_001318821.1 NP_001305750.1 7 1415
NM_001318822.1 NP_001305751.1 5 1230
NM_001318823.1 NP_001305752.1 6 1338
NM_001318824.1 NP_001305753.1 7 1218
NM_182612.3 NP_872418.1 8 1326
NR_134867.1 7 1357
NR_134868.1 8 1420
XM_017017664.1 XP_016873153.1 7 1441
XM_017017667.1 XP_016873156.1 6 1347
XM_017017668.1 XP_016873157.1 6 1429
GenBank mRNA AK091602.1 - 8 1308
AK092816.1 - 8 923
AK094678.1 - 1 257
AK128653.1 - 5 1575
AK293559.1 - 7 1234

View Full Product Details