Gene Transcripts

Gene Symbol
LRRC56
Entrez Gene ID
115399
Gene Name
leucine rich repeat containing 56
Gene Aliases
Chromosome Location
Chr. 11: 518967 - 554916 on Build GRCh38
UniGene ID
Hs.567655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198075.3 NP_932341.1 7, 8 928
XM_005252775.2 XP_005252832.1 7, 8 1752
XM_005252776.2 XP_005252833.1 7, 8 1752
XM_006718132.2 XP_006718195.1 7, 8 1827
XM_006718133.2 XP_006718196.1 6, 7 635
XM_011519875.2 XP_011518177.1 8, 9 1167
XM_011519876.1 XP_011518178.1 8, 9 1823
XM_011519877.2 XP_011518179.1 7, 8 904
XM_017017167.1 XP_016872656.1 8, 9 1242
XM_017017168.1 XP_016872657.1 8, 9 1481
GenBank mRNA AK024495.1 - 6, 7 2395
BC016439.1 - 5, 6 462 3447051
BC035937.1 - 7, 8 914 4816105

View Full Product Details