Gene Transcripts

Gene Symbol
STBD1
Entrez Gene ID
8987
Gene Name
starch binding domain 1
Gene Aliases
GENEX3414, GENX-3414
Chromosome Location
Chr. 4: 76306026 - 76311130 on Build GRCh38
UniGene ID
Hs.109590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003943.4 NP_003934.1 1, 2 964
GenBank mRNA AF062534.1 - 1, 2 347
AK123795.1 - 1, 2 964
BC022301.1 - 1, 2 290 4694574

View Full Product Details