Gene Transcripts
- Gene Symbol
- CHMP2B
- Entrez Gene ID
- 25978
- Gene Name
- charged multivesicular body protein 2B
- Gene Aliases
- ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
- Chromosome Location
- Chr. 3: 87227263 - 87255548 on Build GRCh38
- UniGene ID
- Hs.476930
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001244644.1 |
NP_001231573.1
|
2, 3 |
492 |
|
NM_014043.3 |
NP_054762.2
|
3, 4 |
584 |
|
XM_011533576.2 |
XP_011531878.1
|
3, 4 |
415 |
|
GenBank mRNA |
AF151842.1 |
- |
3, 4 |
543 |
|
AK002165.1 |
- |
3, 4 |
557 |
|
AK002180.1 |
- |
3, 4 |
562 |
|
AK222654.1 |
- |
3, 4 |
544 |
|
AK296072.1 |
- |
2, 3 |
484 |
|
AK316585.1 |
- |
3, 4 |
548 |
|
AL080122.1 |
- |
3, 4 |
557 |
|
BC001553.1 |
- |
3, 4 |
400 |
3460712 |
BI821848.1 |
- |
3, 4 |
561 |
|
BM828410.1 |
- |
3, 4 |
584 |
|
CR533456.1 |
- |
3, 4 |
324 |
|
DA426159.1 |
- |
3, 4 |
555 |
|
View Full Product Details