Gene Transcripts

Gene Symbol
C19orf60
Entrez Gene ID
55049
Gene Name
chromosome 19 open reading frame 60
Gene Aliases
Chromosome Location
Chr. 19: 18588685 - 18592337 on Build GRCh38
UniGene ID
Hs.655317
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100418.1 NP_001093888.1 5 698
NM_001100419.1 NP_001093889.1 5 632
XM_005259977.1 XP_005260034.1 3 619
GenBank mRNA AK000857.1 - 5 586
AK054670.1 - 4 1790
AK091925.1 - 3 2094
AK094710.1 - 1 2285
BC012078.1 - 5 587 4661993
BI836487.1 - 5 509
KP143545.1 - 5 492

View Full Product Details