Gene Transcripts

Gene Symbol
ACSBG2
Entrez Gene ID
81616
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
BGR, BRGL, PRTD-NY3, PRTDNY3
Chromosome Location
Chr. 19: 6135633 - 6193103 on Build GRCh38
UniGene ID
Hs.465720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289177.1 NP_001276106.1 3 511
NM_001289178.1 NP_001276107.1 3 457
NM_001289179.1 NP_001276108.1 3 502
NM_001289180.1 NP_001276109.1 4 342
NM_001321384.1 NP_001308313.1 3 492
NM_030924.4 NP_112186.3 3 502
XM_005259653.3 XP_005259710.1 3 484
XM_017027332.1 XP_016882821.1 3 329
XM_017027333.1 XP_016882822.1 3 484
XM_017027334.1 XP_016882823.1 3 484
GenBank mRNA AJ577571.1 - 3 405
AK093433.1 - 3 470
AK225862.1 - 2 210
AK302604.1 - 3 459
AL136854.1 - 3 243
AY009107.1 - 4 335
AY358766.1 - 3 502
BC022027.1 - 3 433 4825681
BC090046.1 - 3 418 30388900
BI561348.1 - 3 463
DC399703.1 - 4 342
HY011431.1 - 3 489

View Full Product Details