Gene Transcripts

Gene Symbol
AMMECR1L
Entrez Gene ID
83607
Gene Name
AMMECR1 like
Gene Aliases
Chromosome Location
Chr. 2: 127861630 - 127886408 on Build GRCh38
UniGene ID
Hs.546449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199140.1 NP_001186069.1 3 379
NM_031445.2 NP_113633.2 3 351
XM_005263804.4 XP_005263861.1 3 847
XM_005263806.4 XP_005263863.1 3 847
XM_005263807.4 XP_005263864.1 3 847
XM_011511952.2 XP_011510254.1 2 737
XM_011511954.2 XP_011510256.1 3 847
XM_011511956.2 XP_011510258.1 3 847
XM_017005063.1 XP_016860552.1 3 847
GenBank mRNA AK304145.1 - 3 379
BC016181.1 - 3 351 4644159
BC078174.1 - 3 361 6012672

View Full Product Details