Gene Transcripts

Gene Symbol
SELO
Entrez Gene ID
83642
Gene Name
selenoprotein O
Gene Aliases
Chromosome Location
Chr. 22: 50200979 - 50217616 on Build GRCh38
UniGene ID
Hs.365405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031454.1 NP_113642.1 2 653
GenBank mRNA AY324823.1 - 2 653
BC001099.2 - 2 143
BC110866.1 - 2 623 6041782
CR456464.1 - 2 169

View Full Product Details