Gene Transcripts
- Gene Symbol
- CAPN13
- Entrez Gene ID
- 92291
- Gene Name
- calpain 13
- Gene Aliases
- Chromosome Location
- Chr. 2: 30610382 - 30809527 on Build GRCh38
- UniGene ID
- Hs.660911
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144575.2 |
NP_653176.2
|
7, 8 |
955 |
|
XM_011533159.2 |
XP_011531461.1
|
7, 8 |
952 |
|
XM_011533160.2 |
XP_011531462.1
|
8, 9 |
1143 |
|
XM_011533161.2 |
XP_011531463.1
|
7, 8 |
952 |
|
XM_011533163.2 |
XP_011531465.1
|
7, 8 |
952 |
|
XM_017005264.1 |
XP_016860753.1
|
8, 9 |
1143 |
|
XM_017005265.1 |
XP_016860754.1
|
7, 8 |
952 |
|
XM_017005266.1 |
XP_016860755.1
|
7, 8 |
952 |
|
GenBank mRNA |
AK074418.1 |
- |
8, 9 |
1022 |
|
AK290298.1 |
- |
7, 8 |
956 |
|
AK308037.1 |
- |
7, 8 |
932 |
|
BC117345.1 |
- |
7, 8 |
916 |
40125896 |
BC117347.1 |
- |
7, 8 |
916 |
40125898 |
BX640793.1 |
- |
7, 8 |
955 |
|
View Full Product Details