Gene Transcripts

Gene Symbol
SPICE1
Entrez Gene ID
152185
Gene Name
spindle and centriole associated protein 1
Gene Aliases
CCDC52, SPICE
Chromosome Location
Chr. 3: 113442718 - 113515187 on Build GRCh38
UniGene ID
Hs.477144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144718.3 NP_653319.1 4 536
XM_011512461.2 XP_011510763.1 4 293
XM_017005775.1 XP_016861264.1 4 293
XM_017005776.1 XP_016861265.1 4 430
XM_017005777.1 XP_016861266.1 4 353
XM_017005778.1 XP_016861267.1 4 537
GenBank mRNA AK057966.1 - 4 317
AK296160.1 - 4 430
AL832148.1 - 4 505
AL832327.2 - 4 319
AY099107.1 - 4 456
BC036951.1 - 4 477 5500345
BP278430.1 - 4 536

View Full Product Details