Gene Transcripts

Gene Symbol
CXCL5
Entrez Gene ID
6374
Gene Name
C-X-C motif chemokine ligand 5
Gene Aliases
ENA-78, SCYB5
Chromosome Location
Chr. 4: 73995642 - 73998729 on Build GRCh38
UniGene ID
Hs.89714
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002994.4 NP_002985.1 4 1374
GenBank mRNA BC008376.1 - 4 1325

View Full Product Details