Gene Transcripts

Gene Symbol
LYG2
Entrez Gene ID
254773
Gene Name
lysozyme g2
Gene Aliases
LYGH, LYSG2
Chromosome Location
Chr. 2: 99242246 - 99256732 on Build GRCh38
UniGene ID
Hs.436468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175735.3 NP_783862.2 6 665
XM_017003751.1 XP_016859240.1 7 1830
GenBank mRNA BC100882.3 - 5 573
BC100883.2 - 6 634 40003266
BC100884.1 - 5 573 40003273
BC100885.2 - 5 573 40003274

View Full Product Details