Gene Transcripts

Gene Symbol
DAW1
Entrez Gene ID
164781
Gene Name
dynein assembly factor with WD repeats 1
Gene Aliases
ODA16, WDR69
Chromosome Location
Chr. 2: 227871054 - 227924347 on Build GRCh38
UniGene ID
Hs.424594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178821.1 NP_849143.1 3 270
XM_011510755.1 XP_011509057.1 4 879
XM_017003490.1 XP_016858979.1 2 285
GenBank mRNA AK097409.1 - 3 270
AK098821.1 - 3 218
AK127882.1 - 3 827
AK301861.1 - 4 270
BC036377.2 - 3 250 4824989

View Full Product Details