Gene Transcripts
- Gene Symbol
- GEN1
- Entrez Gene ID
- 348654
- Gene Name
- GEN1, Holliday junction 5' flap endonuclease
- Gene Aliases
- Gen
- Chromosome Location
- Chr. 2: 17753687 - 17785920 on Build GRCh38
- UniGene ID
- Hs.467793
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130009.2 |
NP_001123481.2
|
6, 7 |
919 |
|
NM_182625.4 |
NP_872431.4
|
6, 7 |
843 |
|
XM_005262613.4 |
XP_005262670.1
|
6, 7 |
929 |
|
XM_006712005.3 |
XP_006712068.1
|
6, 7 |
826 |
|
XM_011532820.2 |
XP_011531122.1
|
6, 7 |
846 |
|
XM_011532821.2 |
XP_011531123.1
|
6, 7 |
936 |
|
XM_011532822.2 |
XP_011531124.1
|
6, 7 |
816 |
|
GenBank mRNA |
AK131387.1 |
- |
6, 7 |
791 |
|
BC117204.1 |
- |
5, 6 |
741 |
40125755 |
BC117206.1 |
- |
5, 6 |
741 |
40125757 |
CR936783.1 |
- |
6, 7 |
929 |
|
View Full Product Details