Gene Transcripts

Gene Symbol
MTFR1
Entrez Gene ID
9650
Gene Name
mitochondrial fission regulator 1
Gene Aliases
CHPPR, FAM54A2
Chromosome Location
Chr. 8: 65643873 - 65771252 on Build GRCh38
UniGene ID
Hs.584788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145839.1 NP_001139311.1 3 304
NM_014637.3 NP_055452.3 3 304
XM_006716484.2 XP_006716547.2 3 276
XM_011517626.1 XP_011515928.1 3 258
XM_011517627.2 XP_011515929.1 3 194
GenBank mRNA AA015794.1 - 3 236
AK298945.1 - 3 195
AK304716.1 - 3 218
AK316202.1 - 3 229
BC036116.1 - 3 218 5299040
BC043498.2 - 3 203 6057524
BX647507.1 - 3 223
CR456910.1 - 1 35
D13634.1 - 2 52

View Full Product Details