Gene Transcripts

Gene Symbol
DCUN1D4
Entrez Gene ID
23142
Gene Name
defective in cullin neddylation 1 domain containing 4
Gene Aliases
Chromosome Location
Chr. 4: 51843000 - 51916837 on Build GRCh38
UniGene ID
Hs.605388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040402.2 NP_001035492.1 6, 7 665
NM_001287755.1 NP_001274684.1 6, 7 561
NM_001287757.1 NP_001274686.1 7, 8 828
NM_015115.3 NP_055930.2 6, 7 665
XM_005265731.3 XP_005265788.1 6, 7 555
XM_011534379.2 XP_011532681.1 6, 7 1088
XM_011534380.1 XP_011532682.1 6, 7 424
XM_017007912.1 XP_016863401.1 6, 7 518
GenBank mRNA AK027051.1 - 6, 7 484
AK294894.1 - 6, 7 561
AK294896.1 - 7, 8 675
BC053897.1 - 6, 7 531 6173607
BX537944.1 - 6, 7 492
D87466.1 - 6, 7 475
DA770685.1 - 6, 7 555

View Full Product Details