Gene Transcripts

Gene Symbol
MAGEE2
Entrez Gene ID
139599
Gene Name
MAGE family member E2
Gene Aliases
HCA3
Chromosome Location
Chr. X: 75782988 - 75785244 on Build GRCh38
UniGene ID
Hs.356869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138703.4 NP_619648.1 1 308
GenBank mRNA AF490509.1 - 1 290
AK094624.1 - 1 300
BC093964.1 - 1 209 7939809
BC111949.1 - 1 209 8327417
DA793762.1 - 1 308

View Full Product Details