Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199659.2 | NP_001186588.1 | 8 | 1290 | |
NM_001199660.2 | NP_001186589.1 | 8 | 1129 | ||
NM_001286787.1 | NP_001273716.1 | 8 | 1276 | ||
NM_001286819.1 | NP_001273748.1 | 8 | 1361 | ||
NM_144652.3 | NP_653253.1 | 7 | 1146 | ||
XM_006716291.3 | XP_006716354.1 | 8 | 1251 | ||
XM_011544406.1 | XP_011542708.1 | 8 | 1250 | ||
XM_011544407.1 | XP_011542709.1 | 8 | 1212 | ||
XM_011544408.2 | XP_011542710.1 | 8 | 3857 | ||
XM_011544409.2 | XP_011542711.1 | 8 | 1530 | ||
XM_011544410.2 | XP_011542712.1 | 5 | 778 | ||
XM_017013051.1 | XP_016868540.1 | 8 | 1593 | ||
XM_017013052.1 | XP_016868541.1 | 8 | 1321 | ||
XM_017013053.1 | XP_016868542.1 | 8 | 3667 | ||
XM_017013054.1 | XP_016868543.1 | 8 | 1306 | ||
XM_017013055.1 | XP_016868544.1 | 8 | 1449 | ||
XM_017013056.1 | XP_016868545.1 | 8 | 1295 | ||
XM_017013057.1 | XP_016868546.1 | 7 | 3711 | ||
GenBank mRNA | AK058138.1 | - | 7 | 1085 | |
AK302317.1 | - | 8 | 1229 | ||
AK302474.1 | - | 8 | 1101 | ||
AY173945.1 | - | 8 | 1221 | ||
BC029541.1 | - | 5 | 714 | 5297908 | |
BC069830.1 | - | 8 | 1210 |