Gene Transcripts
- Gene Symbol
- IQCE
- Entrez Gene ID
- 23288
- Gene Name
- IQ motif containing E
- Gene Aliases
- 1700028P05Rik
- Chromosome Location
- Chr. 7: 2558972 - 2614734 on Build GRCh38
- UniGene ID
- Hs.520627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC071858.1 |
- |
9, 10 |
893 |
|
View Full Product Details