Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr. X: 101418263 - 101535988 on Build GRCh38
UniGene ID
Hs.729062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256155.2 NP_001243084.2 2 380
NR_028407.2 9 1000
NR_045861.1 6 891
NR_045864.1 6 933
GenBank mRNA AK096955.1 - 6 689
AK126987.1 - 6 754
AK292543.1 - 6 724
BC032236.1 - 6 670
CR933662.1 - 9 975

View Full Product Details