Gene Transcripts

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
Chromosome Location
Chr. X: 14690863 - 14873255 on Build GRCh38
UniGene ID
Hs.554740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018113.2 NP_001018123.1 3 717
NM_001324162.1 NP_001311091.1 3 696
NM_152633.3 NP_689846.1 2 596
XM_011545470.2 XP_011543772.1 3 903
XM_017029355.1 XP_016884844.1 3 795
XM_017029356.1 XP_016884845.1 3 903
GenBank mRNA AK091383.1 - 2 596
BC043596.1 - 3 700
BC136558.1 - 3 699 9020548
BC136560.1 - 2 578 9020550

View Full Product Details