Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
6737
Gene Name
tripartite motif containing 21
Gene Aliases
RNF81, RO52, Ro/SSA, SSA, SSA1
Chromosome Location
Chr. 11: 4384897 - 4393696 on Build GRCh38
UniGene ID
Hs.532357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003141.3 NP_003132.2 2 445
GenBank mRNA AK225564.1 - 2 428
AY742713.1 - 1 332
BC010861.1 - 2 416 3900540
BP280435.1 - 2 445
M34551.1 - 1 370
M62800.1 - 2 417

View Full Product Details