Gene Transcripts

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
Chromosome Location
Chr. 8: 66493256 - 66518524 on Build GRCh38
UniGene ID
Hs.268869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152765.3 NP_689978.2 4 726
GenBank mRNA AK055047.1 - 1 60
AK096336.1 - 5 514
AK308800.1 - 5 553
AK315486.1 - 4 469
BC028400.2 - 4 450 4816822

View Full Product Details