Gene Transcripts

Gene Symbol
SBSPON
Entrez Gene ID
157869
Gene Name
somatomedin B and thrombospondin type 1 domain containing
Gene Aliases
C8orf84, RPESP
Chromosome Location
Chr. 8: 73063077 - 73093272 on Build GRCh38
UniGene ID
Hs.439040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153225.3 NP_694957.3 5 1166
XM_017013145.1 XP_016868634.1 5 1411
GenBank mRNA AK292970.1 - 5 1066
BC042877.1 - 5 1166 5169941

View Full Product Details