Gene Transcripts

Gene Symbol
NSMCE2
Entrez Gene ID
286053
Gene Name
NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase
Gene Aliases
C8orf36, MMS21, NSE2, ZMIZ7
Chromosome Location
Chr. 8: 125091818 - 125367125 on Build GRCh38
UniGene ID
Hs.388297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173685.2 NP_775956.1 8 889
XM_005250875.2 XP_005250932.1 8 897
XM_005250876.4 XP_005250933.1 7 790
XM_011516974.2 XP_011515276.1 9 959
XM_011516975.1 XP_011515277.1 8 854
XM_017013330.1 XP_016868819.1 6 1070
XM_017013331.1 XP_016868820.1 9 927
XM_017013333.1 XP_016868822.1 9 999
GenBank mRNA AK057002.1 - 8 859
BC032797.1 - 8 888 5109691

View Full Product Details