Gene Transcripts

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
Chromosome Location
Chr. 1: 1292384 - 1307925 on Build GRCh38
UniGene ID
Hs.535257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030649.2 NP_085152.2 3, 4 297
XM_005244715.2 XP_005244772.1 3, 4 333
XM_017000233.1 XP_016855722.1 3, 4 248
GenBank mRNA AB051503.1 - 2, 3 1962
AF411981.1 - 1, 2 97
AK093954.1 - 3, 4 344
AK098490.1 - 2, 3 2501
BC051194.1 - 3, 4 297 4819254

View Full Product Details