Gene Transcripts

Gene Symbol
STK11IP
Entrez Gene ID
114790
Gene Name
serine/threonine kinase 11 interacting protein
Gene Aliases
LIP1, LKB1IP, STK11IP1
Chromosome Location
Chr. 2: 219597851 - 219616451 on Build GRCh38
UniGene ID
Hs.22410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052902.3 NP_443134.3 11 1070
XM_011510526.1 XP_011508828.1 11 1050
XM_011510527.2 XP_011508829.1 11 1050
XM_011510529.1 XP_011508831.1 11 1050
GenBank mRNA AB067485.1 - 9 748
AF450267.1 - 11 1024
AK091963.1 - 10 952
AK295609.1 - 10 1238
AK296632.1 - 11 1050
AK300611.1 - 10 957
BC014114.2 - 2 111
BC034051.1 - 11 1047 4819148

View Full Product Details