Gene Transcripts

Gene Symbol
SRP54
Entrez Gene ID
6729
Gene Name
signal recognition particle 54
Gene Aliases
Chromosome Location
Chr. 14: 34982645 - 35029567 on Build GRCh38
UniGene ID
Hs.167535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146282.1 NP_001139754.1 10 1171
NM_003136.3 NP_003127.1 11 1263
XM_005268024.2 XP_005268081.1 11 1516
XM_011537106.1 XP_011535408.1 11 1144
XM_017021615.1 XP_016877104.1 10 1103
GenBank mRNA AK293319.1 - 11 1055
AK300824.1 - 10 1044
AK312853.1 - 11 1208
BC000652.2 - 11 1130 3343314
BC003389.1 - 11 1136 3446599
U51920.1 - 11 983

View Full Product Details