Gene Transcripts

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
Chromosome Location
Chr. 18: 23992773 - 24135952 on Build GRCh38
UniGene ID
Hs.128576
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135993.1 NP_001129465.1 8 1548
NM_001292030.1 NP_001278959.1 3 425
NM_153211.3 NP_694943.2 8 1195
XM_005258198.4 XP_005258255.1 10 1582
XM_017025550.1 XP_016881039.1 10 1676
GenBank mRNA AK091080.1 - 8 1195
AK296442.1 - 3 425
BC032684.1 - 8 997
BC121034.2 - 8 1078 40117554
BC121035.2 - 8 1078 40117556

View Full Product Details