Gene Transcripts
- Gene Symbol
- TTC3
- Entrez Gene ID
- 7267
- Gene Name
- tetratricopeptide repeat domain 3
- Gene Aliases
- DCRR1, RNF105, TPRDIII
- Chromosome Location
- Chr. 21: 37073184 - 37203118 on Build GRCh38
- UniGene ID
- Hs.368214
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001894.1 |
NP_001001894.1
|
3, 4 |
292 |
|
NM_001320703.1 |
NP_001307632.1
|
3, 4 |
498 |
|
NM_001320704.1 |
NP_001307633.1
|
3, 4 |
488 |
|
NM_003316.3 |
NP_003307.3
|
3, 4 |
1583 |
|
XM_005261048.1 |
XP_005261105.1
|
3, 4 |
350 |
|
XM_005261049.1 |
XP_005261106.1
|
3, 4 |
273 |
|
XM_005261050.4 |
XP_005261107.1
|
3, 4 |
3235 |
|
XM_005261052.1 |
XP_005261109.1
|
3, 4 |
273 |
|
XM_005261054.3 |
XP_005261111.1
|
3, 4 |
3227 |
|
XM_011529740.2 |
XP_011528042.1
|
2, 3 |
198 |
|
XM_011529742.2 |
XP_011528044.2
|
3, 4 |
295 |
|
XM_017028458.1 |
XP_016883947.1
|
3, 4 |
289 |
|
XM_017028459.1 |
XP_016883948.1
|
3, 4 |
288 |
|
XM_017028460.1 |
XP_016883949.1
|
3, 4 |
287 |
|
XM_017028461.1 |
XP_016883950.1
|
3, 4 |
350 |
|
XM_017028462.1 |
XP_016883951.1
|
3, 4 |
273 |
|
XM_017028463.1 |
XP_016883952.1
|
3, 4 |
350 |
|
GenBank mRNA |
AK125473.1 |
- |
3, 4 |
2935 |
|
AK291992.1 |
- |
3, 4 |
400 |
|
BC063033.1 |
- |
3, 4 |
310 |
|
BC092466.1 |
- |
3, 4 |
295 |
|
BC137345.1 |
- |
3, 4 |
722 |
9021343 |
D83077.1 |
- |
3, 4 |
1583 |
|
D84294.1 |
- |
2, 3 |
1656 |
|
D84295.1 |
- |
2, 3 |
1656 |
|
View Full Product Details