Gene Transcripts

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
Chromosome Location
Chr. 14: 24205524 - 24208362 on Build GRCh38
UniGene ID
Hs.314432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184739.1 NP_001171668.1 1 356
NM_174944.3 NP_777604.2 1 356
XM_017021226.1 XP_016876715.1 1 552
GenBank mRNA AK131042.1 - 1 356
AY461663.1 - 1 320

View Full Product Details