Gene Transcripts

Gene Symbol
ACBD3
Entrez Gene ID
64746
Gene Name
acyl-CoA binding domain containing 3
Gene Aliases
GCP60, GOCAP1, GOLPH1, PAP7
Chromosome Location
Chr. 1: 226144679 - 226186722 on Build GRCh38
UniGene ID
Hs.520207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022735.3 NP_073572.2 5 901
GenBank mRNA AB043587.2 - 5 909
AK025520.1 - 5 902
AK314468.1 - 5 902
AY150218.1 - 5 880
BC034563.2 - 3 350
BC045533.1 - 5 887 5259930
BC060792.1 - 5 897 4813993

View Full Product Details