Gene Transcripts
- Gene Symbol
- TTC13
- Entrez Gene ID
- 79573
- Gene Name
- tetratricopeptide repeat domain 13
- Gene Aliases
- Chromosome Location
- Chr. 1: 230906241 - 230978872 on Build GRCh38
- UniGene ID
- Hs.424788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024525.4 |
NP_078801.3
|
4, 5 |
566 |
|
XM_005273260.1 |
XP_005273317.1
|
4, 5 |
566 |
|
XM_005273261.2 |
XP_005273318.1
|
4, 5 |
566 |
|
XM_005273264.2 |
XP_005273321.1
|
4, 5 |
566 |
|
XM_011544275.2 |
XP_011542577.1
|
4, 5 |
566 |
|
XM_017002321.1 |
XP_016857810.1
|
4, 5 |
566 |
|
XM_017002322.1 |
XP_016857811.1
|
4, 5 |
566 |
|
XM_017002325.1 |
XP_016857814.1
|
4, 5 |
543 |
|
GenBank mRNA |
AK075386.1 |
- |
4, 5 |
532 |
|
AL832286.1 |
- |
4, 5 |
301 |
|
View Full Product Details