Gene Transcripts

Gene Symbol
CDHR1
Entrez Gene ID
92211
Gene Name
cadherin related family member 1
Gene Aliases
CORD15, PCDH21, PRCAD, RP65
Chromosome Location
Chr. 10: 84194635 - 84219621 on Build GRCh38
UniGene ID
Hs.137556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171971.2 NP_001165442.1 8 882
NM_033100.3 NP_149091.1 8 882
XM_011540337.1 XP_011538639.1 7 1373
XM_011540338.1 XP_011538640.1 7 1373
XM_011540339.1 XP_011538641.1 8 756
XM_011540340.2 XP_011538642.1 7 1373
GenBank mRNA AB053448.1 - 8 755
AB593154.1 - 8 882
AB593155.1 - 8 868
AK124752.1 - 2 136
BC038799.1 - 8 872 5492127

View Full Product Details