Gene Transcripts
- Gene Symbol
- LAPTM4B
- Entrez Gene ID
- 55353
- Gene Name
- lysosomal protein transmembrane 4 beta
- Gene Aliases
- LAPTM4beta, LC27
- Chromosome Location
- Chr. 8: 97775581 - 97852602 on Build GRCh38
- UniGene ID
- Hs.492314
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018407.4 |
NP_060877.3
|
6, 7 |
1027 |
|
GenBank mRNA |
AF317417.1 |
- |
6, 7 |
598 |
|
AF527412.1 |
- |
6, 7 |
956 |
|
AK075326.1 |
- |
6, 7 |
789 |
|
AL136942.1 |
- |
6, 7 |
781 |
|
AY057051.2 |
- |
6, 7 |
1027 |
|
AY261384.1 |
- |
6, 7 |
1046 |
|
BC014129.2 |
- |
6, 7 |
783 |
4581284 |
BC031021.1 |
- |
6, 7 |
808 |
4690368 |
BC038117.1 |
- |
6, 7 |
1575 |
5264567 |
View Full Product Details