Gene Transcripts
- Gene Symbol
- DNAJA4
- Entrez Gene ID
- 55466
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A4
- Gene Aliases
- MST104, MSTP104, PRO1472
- Chromosome Location
- Chr. 15: 78264145 - 78282196 on Build GRCh38
- UniGene ID
- Hs.513053
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130182.1 |
NP_001123654.1
|
5, 6 |
1002 |
|
NM_001130183.1 |
NP_001123655.1
|
5, 6 |
829 |
|
NM_018602.3 |
NP_061072.3
|
6, 7 |
1141 |
|
XM_005254535.3 |
XP_005254592.1
|
6, 7 |
914 |
|
XM_006720601.1 |
XP_006720664.1
|
6, 7 |
916 |
|
GenBank mRNA |
AF173388.1 |
- |
1, 2 |
740 |
|
AK025069.1 |
- |
1 |
124 |
|
AK090898.1 |
- |
2, 3 |
473 |
|
AK096616.1 |
- |
5, 6 |
1002 |
|
AK098079.1 |
- |
6, 7 |
1132 |
|
AL133096.1 |
- |
1 |
914 |
|
AL133433.1 |
- |
1 |
243 |
|
AL133434.1 |
- |
1 |
340 |
|
BC021720.2 |
- |
6, 7 |
1141 |
4831235 |
BC031044.1 |
- |
5, 6 |
864 |
5277834 |
BX648710.1 |
- |
6, 7 |
1023 |
|
View Full Product Details