Gene Transcripts
- Gene Symbol
- BIN3
- Entrez Gene ID
- 55909
- Gene Name
- bridging integrator 3
- Gene Aliases
- Chromosome Location
- Chr. 8: 22620418 - 22669148 on Build GRCh38
- UniGene ID
- Hs.433722
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018688.4 |
NP_061158.1
|
8, 9 |
715 |
|
XM_005273573.3 |
XP_005273630.1
|
7, 8 |
621 |
|
XM_011544586.2 |
XP_011542888.1
|
9, 10 |
800 |
|
GenBank mRNA |
AF271732.1 |
- |
8, 9 |
678 |
|
AK001289.1 |
- |
6, 7 |
577 |
|
AK023980.1 |
- |
8, 9 |
662 |
|
AK124486.1 |
- |
2, 3 |
1000 |
|
AK222470.1 |
- |
8, 9 |
687 |
|
BC001223.1 |
- |
7, 8 |
599 |
3534031 |
BC009824.2 |
- |
8, 9 |
649 |
3953604 |
View Full Product Details