Gene Transcripts
- Gene Symbol
- FAM35A
- Entrez Gene ID
- 54537
- Gene Name
- family with sequence similarity 35 member A
- Gene Aliases
- FAM35A1, bA163M19.1
- Chromosome Location
- Chr. 10: 87094478 - 87191468 on Build GRCh38
- UniGene ID
- Hs.500419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019054.2 |
NP_061927.2
|
5, 6 |
1952 |
|
XM_005269920.3 |
XP_005269977.1
|
5, 6 |
895 |
|
XM_006717903.3 |
XP_006717966.1
|
6, 7 |
2086 |
|
XM_006717905.3 |
XP_006717968.1
|
5, 6 |
2009 |
|
XM_011539876.2 |
XP_011538178.1
|
4, 5 |
1987 |
|
XM_011539877.2 |
XP_011538179.1
|
7, 8 |
2151 |
|
XM_011539878.2 |
XP_011538180.1
|
5, 6 |
2009 |
|
XM_011539879.2 |
XP_011538181.1
|
4, 5 |
1995 |
|
XM_011539881.2 |
XP_011538183.1
|
4, 5 |
1987 |
|
XM_011539882.2 |
XP_011538184.1
|
4, 5 |
1987 |
|
XM_011539883.2 |
XP_011538185.1
|
4, 5 |
940 |
|
XM_017016349.1 |
XP_016871838.1
|
9, 10 |
2395 |
|
XM_017016350.1 |
XP_016871839.1
|
6, 7 |
2354 |
|
XM_017016351.1 |
XP_016871840.1
|
5, 6 |
1942 |
|
XM_017016352.1 |
XP_016871841.1
|
5, 6 |
1942 |
|
XM_017016353.1 |
XP_016871842.1
|
5, 6 |
895 |
|
GenBank mRNA |
AF131775.1 |
- |
2, 3 |
254 |
|
AK022978.1 |
- |
5, 6 |
1952 |
|
AK025606.1 |
- |
3, 4 |
801 |
|
BC051863.2 |
- |
4, 5 |
1898 |
6146593 |
- Gene Symbol
- FAM35BP
- Entrez Gene ID
- 414241
- Gene Name
- family with sequence similarity 35, member A pseudogene
- Gene Aliases
- FAM35B, FAM35EP, bA38L15.1
- Chromosome Location
- Chr. 10: 46610472 - 46651976 on Build GRCh38
- UniGene ID
- Hs.594331
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027632.1 |
|
3, 4 |
1842 |
|
GenBank mRNA |
AW955523.1 |
- |
2, 3 |
412 |
|
- Gene Symbol
- FAM35DP
- Entrez Gene ID
- 439965
- Gene Name
- family with sequence similarity 35, member A pseudogene
- Gene Aliases
- FAM35B2
- Chromosome Location
- Chr. 10: 47689700 - 47731218 on Build GRCh38
- UniGene ID
- Hs.283521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027634.1 |
|
3, 4 |
1842 |
|
GenBank mRNA |
AK294120.1 |
- |
2, 3 |
297 |
|
View Full Product Details