Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300801.1 | NP_001287730.1 | 6 | 958 | |
NM_020845.2 | NP_065896.1 | 6 | 897 | ||
XM_011538590.2 | XP_011536892.1 | 6 | 922 | ||
GenBank mRNA | AB040890.2 | - | 7 | 979 | |
AF334585.1 | - | 5 | 836 | ||
BC141813.1 | - | 6 | 998 |