Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033051.3 | NP_149040.3 | 2 | 1420 | |
GenBank mRNA | AF242557.1 | - | 2 | 1420 | |
AK055571.1 | - | 3 | 1360 | ||
AK314414.1 | - | 2 | 1327 | ||
BC048285.1 | - | 2 | 1272 | 5213610 |