Gene Transcripts
- Gene Symbol
- DMRTC2
- Entrez Gene ID
- 63946
- Gene Name
- DMRT like family C2
- Gene Aliases
- Chromosome Location
- Chr. 19: 41844743 - 41852330 on Build GRCh38
- UniGene ID
- Hs.350507
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK057404.1 |
- |
7 |
1303 |
|
View Full Product Details