Gene Transcripts

Gene Symbol
C18orf8
Entrez Gene ID
29919
Gene Name
chromosome 18 open reading frame 8
Gene Aliases
HsT2591, MIC1, Mic-1
Chromosome Location
Chr. 18: 23503470 - 23531807 on Build GRCh38
UniGene ID
Hs.464779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276342.1 NP_001263271.1 3 350
NM_001318709.1 NP_001305638.1 3 350
NM_013326.4 NP_037458.3 3 350
GenBank mRNA AF143536.1 - 2 97
AK057192.1 - 3 311
AK296230.1 - 3 301
BC002950.1 - 3 275 3538017
BC008305.1 - 3 282 3537084
CN481504.1 - 3 322
DC383182.1 - 3 334

View Full Product Details