Gene Transcripts

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
Chromosome Location
Chr. 1: 215567185 - 215621821 on Build GRCh38
UniGene ID
Hs.335139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319294.1 NP_001306223.1 6, 7 712
NM_001319295.1 NP_001306224.1 6, 7 784
NM_016121.4 NP_057205.2 6, 7 712
XM_005273158.1 XP_005273215.1 6, 7 978
GenBank mRNA AK296275.1 - 4, 5 245
AK316272.1 - 2, 3 113
BC013868.2 - 6, 7 699
BC094879.1 - 6, 7 661 3078971
BC117188.1 - 6, 7 594 40125739
BC126173.1 - 6, 7 594 8991889

View Full Product Details