Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001258006.1 | NP_001244935.1 | 5 | 642 | |
NM_016129.2 | NP_057213.2 | 5 | 642 | ||
XM_011532019.1 | XP_011530321.1 | 5 | 638 | ||
GenBank mRNA | AF100757.1 | - | 5 | 506 | |
AK001148.1 | - | 5 | 638 | ||
AK001210.1 | - | 5 | 469 | ||
AK024005.1 | - | 5 | 527 | ||
AK094238.1 | - | 5 | 1280 | ||
AK223178.1 | - | 5 | 512 | ||
BC004302.1 | - | 5 | 505 | 3623514 | |
BC009292.2 | - | 5 | 505 | 4111685 | |
BC093007.1 | - | 5 | 527 | 30340740 | |
KU178633.1 | - | 5 | 503 | ||
KU178634.1 | - | 5 | 503 |